Learn More
Diagnostic Biosystems CYTOKERATIN CAM52 Cytokeratin (CAM5.2)
Supplier:  Diagnostic Biosystems MOB46901
Encompass_Preferred
Anti-Cytokeratin (CAM 5.2) reagent has a primary reactivity with human keratin proteins that correspond to Moll’s peptides #7 and #8, Mr 48 and 52 kilodaltons (kd), respectively. Cytokeratin 7 and 8 are present on secretory epithelia of normal human tissue but not onstratified squamous epithelium. Anti-Cytokeratin (CAM 5.2) stains most epithelial-derived tissue, including liver, renal tubular epithelium, and hepatocellular and renal cell carcinomas. Anti-Cytokeratin (CAM 5.2) might not react with some squamous cell carcinomas.
Catalog No. 50-238-5682
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.