Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin, LMW Mouse anti-Human, PE-Cyanine7, Clone: KRTL/1377, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254538PECY7

Catalog No. N254538PEC7


Add to cart

Description

Description

Cytokeratin, LMW Monoclonal antibody specifically detects Cytokeratin, LMW in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Cytokeratin, LMW
KRTL/1377
Flow Cytometry
Conjugated, Purified
KRT77
Recombinant human KRT77 protein fragment (exact sequence is proprietary)
Protein A or G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
Cytokeratin LMW, cytokeratin-1B, keratin 77, keratin, type II cytoskeletal 1b, KRT77, type-II keratin Kb39
Mouse
IgG1
0.1 mL
Primary
374454.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit