Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin, LMW Rabbit anti-Human, Clone: KRTL/4440R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen Cytokeratin, LMW
Clone KRTL/4440R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122048
View Documents
Novus Biologicals
NBP30728720UG
20 μg
Each for $159.82
Add to cart
 
NB122047
View Documents
Novus Biologicals
NBP30728710UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

Cytokeratin, LMW Monoclonal specifically detects Cytokeratin, LMW in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Cytokeratin, LMW
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
374454
IgG
Primary
Store at 4°C.
KRTL/4440R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Human
Cytokeratin LMW, cytokeratin-1B, keratin 77, keratin, type II cytoskeletal 1b, KRT77, type-II keratin Kb39
Synthetic peptide corresponding to Cytokeratin, LMW residues within aa100-200 of Cytokeratin, LMW
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit