Learn More
Cytokeratin Mouse anti-Human, Mouse, Rat, Clone: PCK-26, Abnova™
Mouse monoclonal antibody raised against full length native of human cytokeratin.
Manufacturer:  Abnova Corporation MAB12453
Catalog No. 89-934-034
Add to cart
Description
Description
Specifications
Specifications
Cytokeratin
PCK-26
Mouse monoclonal antibody raised against full length native of human cytokeratin.
Lyophilized from ascites fluid (1.2% sodium acetate, 2mg BSA, 0.01mg sodium azide)
Mouse
IgG1
RUO
Primary
Human, Murine, Rat
Immunohistochemistry (PFA fixed), Immunohistochemistry (Frozen), Western Blot
Unconjugated
Immunohistochemistry (1-2 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Lyophilized
Native purified cytokeratin from human epidermis.
100 ug
Store at −20°C. After reconstitution with 1mLof 1.2% sodium acetate or neutral PBS and concentration will be 100μg/mL, store at −20°C. Aliquot to avoid repeated freezing and thawing.
Monoclonal