Learn More
Pan Cytokeratin Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Cytokeratin, Pan Ab-1, Mouse Monoclonal Antibody.
$411.00 - $866.00
Specifications
Antigen | Cytokeratin, Pan Ab-1 |
---|---|
Clone | AE1/AE3 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
MS343PABX
|
Epredia
MS343PABX |
Concentrated | 200 μL |
Each for $866.00
|
|
||||
MS343P1
|
Epredia
MS343P1 |
Concentrated | 500 μL |
Each for $411.00
|
|
||||
MS343P
|
Epredia
MS343P/DEL |
Concentrated | 1 mL |
Each for $643.00
|
|
||||
MS343P1ABX
|
Epredia
MS343P1ABX |
Concentrated | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS343R1
|
Epredia
MS343R1 |
Prediluted | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS343R7
|
Epredia
MS343R7 |
Prediluted | 7 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS343RQ
|
Epredia
MS343RQ |
Ready-to-Use | 12 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI <5.7) and basic (pI >6.0) subfamilies. The acidic keratins have molecular weights of 56.5, 55, 51, 50, 50', 48, 46, 45, and 40 kDa. The basic keratins have molecular weights of 65-67, 64, 59, 58, 56 and 52 kDa. Members of acidic and basic subfamilies are found together in pairs. The composition of keratin pairs varies with cell type, differentiation status and environment. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.Host Species: Mouse
Clone: AE1/AE3
Isotype: IgG1 + IgG1
Species Reactivity: Human, Monkey, Cow, Rabbit, Mouse, Rat and Chicken. Others not known.
Immunogen: Human epidermal keratin
Positive Control: Skin, Lung CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifications
Cytokeratin, Pan Ab-1 | |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Human epidermal keratin |
AE1/AE3 | |
Monoclonal | |
Mouse | |
Cytoskeletal Protein | |
Primary |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.