Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin, pan Mouse anti-Human, Mouse, Rat, Porcine, Several Species, Alexa Fluor™ 750, Clone: C11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP234572AF750

Catalog No. NP23472A70


Add to cart

Description

Description

Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Guinea Pig, Goat, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications

Specifications

Cytokeratin, pan
C11
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Conjugated, Purified
Mouse
IgG1
0.1 mL
Primary
3848
Amphibian, Bovine, Goat, Guinea Pig, Human, Mouse, Porcine, Primate, Rat
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
KRT1
Keratin-enriched preparation from cultured human A431
Protein G purified
Store at 4°C in the dark.
Monoclonal
This MAb recognizes cytokeratin 4, 5, 6, 8, 10, 13, and 18. This is a broad-spectrum antibody, which has been reported to differentiate epithelial tumors from non-epithelial tumors. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit