Learn More
Invitrogen™ Cytokeratin Pan Type I Monoclonal Antibody (AE1)

Description
MA1-5761 detects most human acidic (type I) keratins K10, K14, K15, K16, K19. MA1-5761 has been successfully used in Western blot, immunomicroscopy and immunohistochemistry (paraffin) procedures. For Western blotting, do not use milk for blocking nonspecific binding because it contains some keratins (bovine serum albumin is recommended). The MA1-5761 immunogen is human epidermal keratins.
Specifications
Specifications
| Antigen | Cytokeratin Pan Type I |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | AE1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | KRT16 |
| Gene Accession No. | O93256, P02533, P02535, P05783, P05784, P05785, P06394, P08727, P08728, P08779, P13645, P19001, P19012, P35527, Q5BJY9, Q61414, Q61781, Q63279, Q6IFV1, Q6IFV3, Q6IFW6, Q6PVZ1, Q6RHW0, Q8CIS9, Q9Z2K1 |
| Gene Alias | 40-kDa keratin intermediate filament; 56 kDa cytokeratin; 67 kDa cytokeratin; AA960620; Acidic Cytokeratin; AI324768; AI528832; AI626930; AI663979; AL022697; alpha keratin; AU019895; BCIE; BIE; Card2; Cell proliferation-inducing gene 46 protein; CK1; CK-1; CK10; CK-10; CK14; CK-14; CK15; CK-15; CK16; CK-16; CK18; CK-18; CK19; CK-19; CK8; CK-8; CK-9; CYK18; CYK8; CYKER; cytokeratin 1; cytokeratin 10; cytokeratin 14; cytokeratin 15; cytokeratin 16; cytokeratin 18; cytokeratin 19; cytokeratin 8; cytokeratin 8 (370 AA); cytokeratin 9; cytokeratin endo A; Cytokeratin endo B; cytokeratin VIB; cytokeratin VII; Cytokeratin-1; Cytokeratin-10; cytokeratin-14; Cytokeratin-15; cytokeratin-16; Cytokeratin-18; cytokeratin-19; Cytokeratin-6B; cytokeratin-7; cytokeratin-8; Cytokeratin-9; cytokeratin-A; D130054E02Rik; EBS3; EBS4; EGK_03685; EHK; EHK1; Endo B; EndoA; EndoC; epidermal keratin 10; epidermal keratin VII; epidermolysis bullosa simplex, Dowling-Meara, Koebner; epidermolytic hyperkeratosis 1; epidermolytic hyperkeratosis; keratosis palmaris et plantaris; epithelial keratin 1; epithelial keratin 10; Epithelial keratin-1; EPPK; FNEPPK; focal non-epidermolytic palmoplantar keratoderma; GK-19; Hair alpha protein; HMWCK; intermediate filament protein; K1; K10; K14; K15; K16; K18; K19; K1C1; K1CO; K1CP; K1CS; K2C8; K8; K9; Ka10; Ka14; Ka15; Ka16; Ka19; Kb1; KER1; Ker10; KERA; keratin 1; keratin 1 (epidermolytic hyperkeratosis); keratin 1, type II; keratin 10; keratin 10 (epidermolytic hyperkeratosis); keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris); keratin 10, type I; keratin 13; keratin 14; keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner); keratin 14, type I; keratin 15; keratin 15, type I; keratin 16; keratin 16 (focal non-epidermolytic palmoplantar keratoderma); keratin 16, type I; keratin 18; keratin 18, type I; keratin 19; keratin 19, type I; keratin 24; keratin 8; keratin 8, type II; keratin 86; keratin 9; keratin 9 (epidermolytic palmoplantar keratoderma); keratin 9, type I; keratin complex 1, acidic, gene 10; keratin complex 1, acidic, gene 14; keratin complex 1, acidic, gene 15; keratin complex 1, acidic, gene 16; keratin complex 1, acidic, gene 18; keratin complex 1, acidic, gene 19; keratin complex 1, acidic, gene 9; keratin complex 2, basic, gene 1; keratin complex 2, basic, gene 8; keratin D; keratin intermediate filament 16a; keratin intermediate filament 16b; keratin, type I cytoskeletal 10; keratin, type I cytoskeletal 10-like; keratin, type I cytoskeletal 13; keratin, type I cytoskeletal 14; keratin, type I cytoskeletal 14; keratin, type I cytoskeletal 16; keratin, type I cytoskeletal 15; Keratin, type I cytoskeletal 16; keratin, type I cytoskeletal 18; Keratin, type I cytoskeletal 19; keratin, type I cytoskeletal 59 kDa; Keratin, type I cytoskeletal 9; keratin, type I, 40-kd; keratin, type II cytoskeletal 1; Keratin, type II cytoskeletal 8; keratin-1; Keratin-10; keratin-14; Keratin-15; keratin-15, basic; keratin-15, beta; keratin-16; Keratin-18; keratin-19; Keratin-8; keratin-9; Kerd; KO; KPP; KRT1; krt1 {ECO:0000250; Krt-1.10; Krt-1.14; Krt-1.19; KRT10; krt10 {ECO:0000250; Krt1-10; Krt1-14; Krt1-15; Krt1-16; Krt1-18; Krt1-19; KRT13; KRT14; Krt15; krt15 {ECO:0000250; Krt16; KRT16A; KRT18; krt18 {ECO:0000250; KRT19; Krt1-9; KRT1A; Krt-2.1; Krt-2.8; Krt2-1; KRT24; Krt2-8; Krt8; Krt86; krt9; KRTA; KRTB; LOW QUALITY PROTEIN: keratin, type I cytoskeletal 15; NEPPK; NFJ; pan keratin; pankeratin; PC1; PIG46; Spermatid perinuclear ring manchette protein K9; suprabasal cytokeratin 10; type I alpha-keratin 14; type I alpha-keratin 15; type I cytoskeletal 15; type I cytoskeletal 9; type I keratin KA10; Type I keratin Ka14; type I keratin KA15; type I keratin KA16; Type I keratin Ka19; type II cytokeratin A (no.8); type II keratin Kb1; type-II keratin Kb1; Type-II keratin Kb8; UniProtKB:P04264}; UniProtKB:P05783}; UniProtKB:P13645}; UniProtKB:P19012}; unnamed protein product |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.