Learn More
Invitrogen™ Cytokeratin Pan Type II Monoclonal Antibody (AE3)

Description
MA1-5762 reacts with all basic (type II) human keratins in human samples, including K1, K2, K3, K4, K5, K6, K7 and K8. MA1-5762 has been successfully used in Western blot, immunohistochemistry and electron microscopy procedures. The MA1-5762 immunogen is human epidermal keratins.
Specifications
Specifications
| Antigen | Cytokeratin Pan Type II |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | AE3 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | KRT7 |
| Gene Accession No. | O93532, P02538, P04104, P04264, P05786, P05787, P07744, P08729, P11679, P12035, P13647, P19013, P35908, P50446, Q01546, Q10758, Q148H8, Q14CN4, Q29426, Q29S21, Q3TTY5, Q3UV17, Q4FZU2, Q5XQN5, Q6IG00, Q6IG02, Q6IG04, Q6IG12, Q6IME9, Q6IMF3, Q6P6Q2, Q922U2, Q9DCV7 |
| Gene Alias | 2310016L08Rik; 3300001P10Rik; 57kd keratin; 57kDa keratin; 58 kda cytokeratin; 60-kDa keratin; 65 kDa cytokeratin; 67 kDa cytokeratin; AA960620; AI507495; AL022697; alpha keratin; AU019895; AW108092; AW146334; BB005427; Card2; CK 2e; CK1; CK-1; CK-2e; CK3; CK-3; CK4; CK-4; CK5; CK-5; CK6A; CK-6A; CK6C; CK-6C; CK6D; CK-6D; CK-6E; CK7; CK-7; CK-72; CK8; CK-8; CYK4; CYK8; CYKER; cytokeratin 1; cytokeratin 3; cytokeratin 4; cytokeratin 5; cytokeratin 6A; cytokeratin 6C; cytokeratin 6D; cytokeratin 7; cytokeratin 8; cytokeratin 8 (370 AA); cytokeratin endo A; Cytokeratin otokeratin; cytokeratin type II, component Ib/c; cytokeratin type II, component III; Cytokeratin-1; Cytokeratin-2e; cytokeratin-3; Cytokeratin-4; cytokeratin-5; Cytokeratin-6A; cytokeratin-6C; Cytokeratin-6D; cytokeratin-6E; cytokeratin-7; Cytokeratin-72; cytokeratin-8; Cytokeratin-A; cytoskeletal 57 kDa keratin; D15Wsu77e; DDD; DDD1; ear specific cytokeratin; EBS2; EGK_03684; EGK_03685; EHK; EHK1; EndoA; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; epidermolytic hyperkeratosis 1; epithelial keratin 1; epithelial keratin 2e; Epithelial keratin-1; Epithelial keratin-2e; EPPK; Hair alpha protein; Hom s 5; K1; K2C7; K2C8; K2E; K3; K3 keratin; K4; K5; K6A; K6a keratin; K6C; K6D; K6irs; K6IRS2; K7; K72; K8; kamp-keratin derived antimicrobial peptide; Kb1; Kb2; Kb35; Kb4; Kb7; KDAMP; KER1; Ker2; KERA; keratin; keratin 1; keratin 1 (epidermolytic hyperkeratosis); keratin 1, type II; keratin 2; keratin 2 (epidermal ichthyosis bullosa of Siemens); keratin 2 epidermis; keratin 2, type II; keratin 2A; keratin 2A (epidermal ichthyosis bullosa of Siemens); keratin 3; keratin 3, type II; keratin 4; keratin 4, type II; keratin 5; keratin 5 (epidermolysis bullosa simplex Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 6 alpha; keratin 6, inner root sheath, 2; keratin 6A; keratin 6A, , type II; keratin 6A, type II; keratin 7; keratin 7, type II; keratin 72; keratin 72, pseudogene; keratin 72, type II; keratin 8; keratin 8, type II; keratin 86; keratin complex 2, basic, gene 1; keratin complex 2, basic, gene 17; keratin complex 2, basic, gene 35; keratin complex 2, basic, gene 4; keratin complex 2, basic, gene 5; keratin complex 2, basic, gene 6a; keratin complex 2, basic, gene 6c; keratin complex 2, basic, gene 7; keratin complex 2, basic, gene 8; keratin complex 2, gene 17; keratin complex 2, gene 6a; keratin IV; keratin K3; keratin K6; keratin K6h; keratin protein K6irs; keratin, 55K type II cytoskeletal; keratin, epidermal type II, K6A; keratin, simple epithelial type I, K7; keratin, type II cytoskeletal 1; Keratin, type II cytoskeletal 2 epidermal; keratin, type II cytoskeletal 3; Keratin, type II cytoskeletal 4; keratin, type II cytoskeletal 5; keratin, type II cytoskeletal 59 kDa, component IV; keratin, type II cytoskeletal 60 kDa, component III; keratin, type II cytoskeletal 6A; keratin, type II cytoskeletal 6C; keratin, type II cytoskeletal 7; Keratin, type II cytoskeletal 72; keratin, type II cytoskeletal 8; Keratin, type II cytoskeletal cochleal; keratin-1; Keratin-2 epidermis; keratin-2e; Keratin-3; Keratin-4; keratin-5; keratin-6A; Keratin-6-alpha; keratin-6C; keratin-7; Keratin-72; keratin-8; KERIB; KO; Krt1; krt1 {ECO:0000250; KRT1A; Krt2; krt2 {ECO:0000250; Krt-2.1; Krt-2.4; Krt-2.8; Krt2-1; Krt2-17; Krt2-2; Krt2-35; Krt2-4; Krt2-5; Krt2-6; Krt2-6a; Krt2-6c; Krt2-7; KRT2-8; KRT2A; KRT2E; KRT3; KRT4; krt4 {ECO:0000250; Krt5; KRT5A; KRT6; Krt6a; KRT6C; KRT6D; KRT6IRS2; Krt7; krt7 {ECO:0000250; Krt72; Krt72-ps; Krt8; Krt86; KRTA; KRTE; LOC683313; LOW QUALITY PROTEIN: keratin, type II cytoskeletal 7; MGC116967; MGC116968; mK6[a]; MK6a; mK6-alpha; NEPPK; otokeratin; pan keratin; PC3; Sak57; sarcolectin; SCL; similar to keratin 7; similar to keratin complex 2, basic, gene 6a; Tfip8; tuftelin interacting protein 8 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.