Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin Type I Monoclonal Antibody (AE-1)
GREENER_CHOICE

Catalog No. 01671422
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-422 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-422 Supplier Invitrogen™ Supplier No. MA135367
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin pan is part of a subfamily of intermediate filament proteins that are characterized by remarkable biochemical diversity, and represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight between 40 kDa- 68 kDa, and an isoelectric pH between 4.9-7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. The composition of cytokeratin pairs vary with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

Cytokeratin pan is part of a subfamily of intermediate filament proteins that are characterized by remarkable biochemical diversity, and represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight between 40 kDa- 68 kDa, and an isoelectric pH between 4.9-7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. The composition of cytokeratin pairs vary with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin Type I
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone AE-1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene KRT16
Gene Accession No. O93256, P02533, P02535, P05783, P05784, P05785, P06394, P08727, P08728, P08779, P13645, P19001, P19012, P35527, Q5BJY9, Q61414, Q61781, Q63279, Q6IFV1, Q6IFV3, Q6IFW6, Q6PVZ1, Q6RHW0, Q8CIS9, Q9Z2K1
Gene Alias 40-kDa keratin intermediate filament; 56 kDa cytokeratin; 67 kDa cytokeratin; AA960620; Acidic Cytokeratin; AI324768; AI528832; AI626930; AI663979; AL022697; alpha keratin; AU019895; BCIE; BIE; Card2; Cell proliferation-inducing gene 46 protein; CK1; CK-1; CK10; CK-10; CK14; CK-14; CK15; CK-15; CK16; CK-16; CK18; CK-18; CK19; CK-19; CK8; CK-8; CK-9; CYK18; CYK8; CYKER; cytokeratin 1; cytokeratin 10; cytokeratin 14; cytokeratin 15; cytokeratin 16; cytokeratin 18; cytokeratin 19; cytokeratin 8; cytokeratin 8 (370 AA); cytokeratin 9; cytokeratin endo A; Cytokeratin endo B; cytokeratin VIB; cytokeratin VII; Cytokeratin-1; Cytokeratin-10; cytokeratin-14; Cytokeratin-15; cytokeratin-16; Cytokeratin-18; cytokeratin-19; Cytokeratin-6B; cytokeratin-7; cytokeratin-8; Cytokeratin-9; cytokeratin-A; D130054E02Rik; EBS3; EBS4; EGK_03685; EHK; EHK1; Endo B; EndoA; EndoC; epidermal keratin 10; epidermal keratin VII; epidermolysis bullosa simplex, Dowling-Meara, Koebner; epidermolytic hyperkeratosis 1; epidermolytic hyperkeratosis; keratosis palmaris et plantaris; epithelial keratin 1; epithelial keratin 10; Epithelial keratin-1; EPPK; FNEPPK; focal non-epidermolytic palmoplantar keratoderma; GK-19; Hair alpha protein; HMWCK; intermediate filament protein; K1; K10; K14; K15; K16; K18; K19; K1C1; K1CO; K1CP; K1CS; K2C8; K8; K9; Ka10; Ka14; Ka15; Ka16; Ka19; Kb1; KER1; Ker10; KERA; keratin 1; keratin 1 (epidermolytic hyperkeratosis); keratin 1, type II; keratin 10; keratin 10 (epidermolytic hyperkeratosis); keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris); keratin 10, type I; keratin 13; keratin 14; keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner); keratin 14, type I; keratin 15; keratin 15, type I; keratin 16; keratin 16 (focal non-epidermolytic palmoplantar keratoderma); keratin 16, type I; keratin 18; keratin 18, type I; keratin 19; keratin 19, type I; keratin 24; keratin 8; keratin 8, type II; keratin 86; keratin 9; keratin 9 (epidermolytic palmoplantar keratoderma); keratin 9, type I; keratin complex 1, acidic, gene 10; keratin complex 1, acidic, gene 14; keratin complex 1, acidic, gene 15; keratin complex 1, acidic, gene 16; keratin complex 1, acidic, gene 18; keratin complex 1, acidic, gene 19; keratin complex 1, acidic, gene 9; keratin complex 2, basic, gene 1; keratin complex 2, basic, gene 8; keratin D; keratin intermediate filament 16a; keratin intermediate filament 16b; keratin, type I cytoskeletal 10; keratin, type I cytoskeletal 10-like; keratin, type I cytoskeletal 13; keratin, type I cytoskeletal 14; keratin, type I cytoskeletal 14; keratin, type I cytoskeletal 16; keratin, type I cytoskeletal 15; Keratin, type I cytoskeletal 16; keratin, type I cytoskeletal 18; Keratin, type I cytoskeletal 19; keratin, type I cytoskeletal 59 kDa; Keratin, type I cytoskeletal 9; keratin, type I, 40-kd; keratin, type II cytoskeletal 1; Keratin, type II cytoskeletal 8; keratin-1; Keratin-10; keratin-14; Keratin-15; keratin-15, basic; keratin-15, beta; keratin-16; Keratin-18; keratin-19; Keratin-8; keratin-9; Kerd; KO; KPP; KRT1; krt1 {ECO:0000250; Krt-1.10; Krt-1.14; Krt-1.19; KRT10; krt10 {ECO:0000250; Krt1-10; Krt1-14; Krt1-15; Krt1-16; Krt1-18; Krt1-19; KRT13; KRT14; Krt15; krt15 {ECO:0000250; Krt16; KRT16A; KRT18; krt18 {ECO:0000250; KRT19; Krt1-9; KRT1A; Krt-2.1; Krt-2.8; Krt2-1; KRT24; Krt2-8; Krt8; Krt86; krt9; KRTA; KRTB; LOW QUALITY PROTEIN: keratin, type I cytoskeletal 15; NEPPK; NFJ; pan keratin; pankeratin; PC1; PIG46; Spermatid perinuclear ring manchette protein K9; suprabasal cytokeratin 10; type I alpha-keratin 14; type I alpha-keratin 15; type I cytoskeletal 15; type I cytoskeletal 9; type I keratin KA10; Type I keratin Ka14; type I keratin KA15; type I keratin KA16; Type I keratin Ka19; type II cytokeratin A (no.8); type II keratin Kb1; type-II keratin Kb1; Type-II keratin Kb8; UniProtKB:P04264}; UniProtKB:P05783}; UniProtKB:P13645}; UniProtKB:P19012}; unnamed protein product
Gene Symbols KRT10, Krt14, KRT15, KRT16, KRT18, KRT19, KRT9
Host Species Mouse
Immunogen Human epidermal keratin.
Purification Method purified
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009029, 100138169, 100341120, 100341376, 100354174, 101749333, 107656, 16661, 16664, 16665, 16666, 16668, 16669, 266717, 281888, 287700, 287701, 294853, 303530, 360626, 3857, 3858, 3861, 3866, 3868, 3875, 3880, 395861, 404111, 408039, 408040, 450225, 506480, 507464, 514812, 771977, 788727
Target Species Human, Mouse, Rat, Rabbit, Bovine, Chicken
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.