Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin17/36 Rabbit anti-Human, Clone: SR45-06, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267318
Description
Cytokeratin 17/36 Monoclonal specifically detects Cytokeratin 17/36 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Cytokeratin17/36 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:50-1:400 | |
39.1, CK-17, cytokeratin-17, K17, keratin 17, keratin, type I cytoskeletal 17, keratin-17, PC, PC2, PCHC1 | |
Rabbit | |
Protein A purified | |
Cancer | |
3872 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
SR45-06 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
KRT17 | |
Synthetic peptide within Human Cytokeratin 17/36 aa 1-50/ 432. (SwissProt: Q04695 Human; SwissProt: Q9QWL7 Mouse; SwissProt: Q6IFU8 Rat) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction