Learn More
Cytokeratin5/8 Mouse anti-Canine, Janelia Fluor 646, Clone: RCK102, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP197730JF646
Catalog No. NBP97730646
Add to Cart
Description
Description
Cytokeratin 5/8 Monoclonal specifically detects Cytokeratin 5/8 in Human, Mouse, Rat, Porcine, Canine, Feline, Hamster, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
Cytokeratin5/8
Monoclonal
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
KRT5
Clone RCK102 is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0 mouse myeloma cells with spleen cells from a mouse immunized with cytokeratins from a human lung cancer cell line (MR21).
0.1 mL
3852
Canine, Feline, Hamster, Human, Mouse, Porcine, Rabbit, Rat
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
RCK102
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
cell proliferation-inducing protein 46, CK18, CK5, CYK18, DDD, K18, K1CO, K5, Keratin, Keratin 18, Keratin 5, KRT18, krt5, KRT5A, KRTB, type I cytoskeletal 18, type II cytoskeletal 5
Mouse
Protein A or G purified
Primary
RCK102 is reacting with cytokeratin 5 and cytokeratin 8. This monoclonal antibody recognizes virtually all epithelial tissues and carcinomas.
Store at 4°C in the dark.
IgG1