Learn More
Cytomegalovirus Goat anti-Cytomegalovirus, Polyclonal, Abnova™
Goat Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29837
Catalog No. 89-033-682
Add to cart
Description
Description
Goat polyclonal antibody raised against cytomegalovirus (CMV).
  • High specificity
  • High reactivity
Specifications
Specifications
Cytomegalovirus
Unconjugated
ELISA Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Liquid
Purified cytomegalovirus strain AD169.
RUO
Primary
Cytomegalovirus
ELISA, Immunofluorescence, Western Blot
Goat polyclonal antibody raised against cytomegalovirus (CMV).
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Goat
500 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)