Learn More
Cytomegalovirus ICP8 Mouse anti-Cytomegalovirus, Clone: P1209, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13524
Catalog No. 89-033-487
Add to cart
Description
Description
Mouse monoclonal antibody raised against cytomegalovirus (CMV) ICP8.
  • High specificity
  • High reactivity
Specifications
Specifications
Cytomegalovirus ICP8
P1209
Mouse monoclonal antibody raised against cytomegalovirus (CMV) ICP8.
In PBS, pH 7.4.
Mouse
Protein G purification
RUO
Primary
This antibody reacts with ICP8 of cytomegalovirus.
Immunofluorescence, Western Blot
Unconjugated
Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Liquid
IgG
500 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Cytomegalovirus
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)