Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytomegalovirus Antibody (HCMV16 (51C1)) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
$204.00 - $482.50
Specifications
Antigen | Cytomegalovirus |
---|---|
Clone | HCMV16 (51C1) |
Concentration | 1.0 mg/mL |
Dilution | ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry (Frozen), Immunofluorescence, Immunohistochemistry, Immunoprecipitation, ELISA, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017574
|
Novus Biologicals
NB100643980.025MG |
0.025 mg |
Each for $204.00
|
|
|||||
NB10064398
|
Novus Biologicals
NB10064398 |
0.1 mg |
Each for $482.50
|
|
|||||
Description
Cytomegalovirus Monoclonal specifically detects Cytomegalovirus in Cytomegalovirus, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
Cytomegalovirus | |
1.0 mg/mL | |
Immunohistochemistry (Frozen), Immunofluorescence, Immunohistochemistry, Immunoprecipitation, ELISA, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Virus, Cytomegalovirus (CMV) | |
Cytomegalovirus extracts | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
HCMV16 (51C1) | |
ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
CMV; HHV 5; HHV5; Human herpesvirus 5 | |
IgG1 | |
Protein A purified | |
NB100-64398 is specific for Cytomegalovirus glycoprotein H (gH) late neutralizing antigen and reacts with MRC-5 infected cells. It does not react with HSV, VZV, EBV or uninfected cells. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title