Learn More
R&D Systems™ Cytonin™
R&D Systems™ Cytonin™ offers an alternative to Proteinase K for permeabilizing cells and tissues prior to labeling using any of the TACS™ In Situ Apoptosis Detection kits.
Manufacturer:  R&D Systems™ 48760501
Catalog No. 48-760-501
Add to cart
This item is not returnable. View return policy
Description
Description
  • Protease free, saponin-based buffer designed specifically for in situdetection of apoptosis. Cytonin™ should be used when protease treatment must be avoided. For example, it may be used for tissues with low cellularity or little connective tissue that do not withstand protease treatment. Neuropore is recommended for double labeling experiments for the detection of DNA fragmentation in conjunction with immunohistochemistry on antigens that are sensitive to proteolysis.
Specifications
Specifications
6mL
Store the unopened product at 2 - 8° C. Do not use past expiration date.