Learn More
R&D Systems™ Cytonin
Use in dual-labeling experiments detecting both DNA fragmentation and antigens that may be sensitive to proteinase K treatment. Cytonin can be used to permeabilize cells and tissues prior to labeling with TACS™ in situ Apoptosis Detection Kits.
Manufacturer:  R&D Systems™ 48760502
Catalog No. 1087086
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Non-proteolytic, saponin-based permeabilization reagent designed specifically for in situ detection of apoptosis
  • Use when protease treatment must be avoided
Specifications
Specifications
2 x 5mL
Store the unopened product at 2 - 8° C. Do not use past expiration date.