Learn More
Cell Biolabs IncSupplier Diversity Partner CYTOSELECT 24W INVASION KT ECM  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner CBA110T
The CytoSelect 24-Well Cell Invasion Assay Kit (Trial Size) contains 4 membrane inserts coated with a uniform layer of dried basement membrane matrix. Cells degrade the matrix layer and pass thru the membrane. Colorimetric readout.
Catalog No. 50-672-643