Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cytosine Deaminase Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA185365
Description
In Western blot, this antibody detects a band at ~13 and 17 kDa. Sheep anti cytosine deaminase antibody recognizes cytosine deaminase, also known as cytosine aminohydrolase.

Specifications
Cytosine Deaminase | |
Polyclonal | |
Unconjugated | |
FCY1 | |
Cytosine aminohydrolase; Cytosine deaminase; FCY1; YP9499.17; YPR062W | |
Sheep | |
Protein G | |
RUO | |
856175 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
5 mg/mL | |
PBS with 0.09% sodium azide | |
Q12178 | |
FCY1 | |
Purified yeast cytosine deaminase. | |
1 mL | |
Primary | |
Yeast | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction