Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL Mouse anti-Human, Alexa Fluor™ 750, Clone: 1500000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP222557AF750

Catalog No. NP2227A70


Add to cart

Description

Description

Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL Monoclonal antibody specifically detects Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL in Human samples. It is validated for Western Blot,ELISA,Immunocytochemistry,Immunofluorescence.
Specifications

Specifications

Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL
1500000
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
CN2cytosolic non-specific dipeptidase, CNDP dipeptidase 2, CNDP dipeptidase 2 (metallopeptidase M20 family), CPGLcytosolic nonspecific dipeptidase, FLJ10830, HsT2298, PEPAEC 3.4.13.18, Peptidase AGlutamate carboxypeptidase-like protein 1
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CNDP2
Recombinant human CNDP2 (1-475aa) purified from E. coli
0.1 mL
Primary
55748
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit