Learn More
Cytosolic beta-Glucosidase/GBA3 Rabbit anti-Human, Alexa Fluor 594, Clone: 8, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP289924AF594
Catalog No. NB110109
Add to cart
Description
Description
Cytosolic beta-Glucosidase/GBA3 Monoclonal specifically detects Cytosolic beta-Glucosidase/GBA3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Cytosolic beta-Glucosidase/GBA3
8
50 mM Sodium Borate
CBG, CBGL1GLUC, cytosolic beta-glucosidase, Cytosolic beta-glucosidase-like protein 1, EC 3.2.1.21, glucosidase, beta, acid 3 (cytosolic), klotho-related protein, KLrP, MGC104276, MGC126878
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cytosolic beta-Glucosidase/GBA3 (NP_066024.1; Met1-Leu469).
Protein A purified
RUO
Primary
57733
Flow Cytometry, Immunofluorescence
Alexa Fluor 594
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human