Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytosolic Sulfotransferase 1A1/SULT1A1 Mouse anti-Human, Allophycocyanin/Cy7, Clone: OTI9B7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271308APCCY7

Catalog No. N271308APC7


Add to cart

Description

Description

Cytosolic Sulfotransferase 1A1/SULT1A1 Monoclonal antibody specifically detects Cytosolic Sulfotransferase 1A1/SULT1A1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Cytosolic Sulfotransferase 1A1/SULT1A1
OTI9B7
Flow Cytometry
Affinity Purified, Conjugated
SULT1A1
Full length recombinant protein of human Cytosolic Sulfotransferase 1A1/SULT1A1 (NP_001046) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
Aryl sulfotransferase 1, EC 2.8.2, EC 2.8.2.1, HAST1/HAST2, Phenol sulfotransferase 1, Phenol-sulfating phenol sulfotransferase 1, P-PST, P-PST 1, PST, ST1A1, ST1A3, STP1MGC5163, STPMGC131921, sulfotransferase 1A1, sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1, Thermostable phenol sulfotransferase, thermostable phenol sulfotransferase1, ts-PST, TSPST1
Mouse
IgG1
0.1 mL
Primary
6817.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit