Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytosolic Sulfotransferase 1B1/SULT1B1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP154829

 View more versions of this product

Catalog No. NBP154829


Add to cart

Description

Description

Cytosolic Sulfotransferase 1B1/SULT1B1 Polyclonal antibody specifically detects Cytosolic Sulfotransferase 1B1/SULT1B1 in Human, Rat, Porcine, Bovine, Canine samples. It is validated for Western Blot.
Specifications

Specifications

Cytosolic Sulfotransferase 1B1/SULT1B1
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
SULT1B1
Synthetic peptides corresponding to SULT1B1(sulfotransferase family, cytosolic, 1B, member 1) The peptide sequence was selected from the N terminal of SULT1B1. Peptide sequence MLSPKDILRKDLKLVHGYPMTCAFASNWEKIEQFHSRPDDIVIATYPKSG.
35 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Canine: 86%; Rat: 86%; Porcine: 79%.
Bovine, Canine, Human, Porcine, Rat
Western Blot
Western Blot 1:100-1:2000
O43704
EC 2.8.2, EC 2.8.2.-, EC 2.8.2.1, EC 2.8.2.4, MGC13356, ST1B1, ST1B2sulfotransferase family cytosolic 1B member 1, Sulfotransferase 1B1, Sulfotransferase 1B2, sulfotransferase family, cytosolic, 1B, member 1, SULT1B2, Thyroid hormone sulfotransferase
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
27284
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit