Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytosolic Sulfotransferase 1C2/SULT1C2 Mouse anti-Human, PE-Cyanine7, Clone: OTI4G1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271313PECY7

Catalog No. N271313PEC7


Add to cart

Description

Description

Cytosolic Sulfotransferase 1C2/SULT1C2 Monoclonal antibody specifically detects Cytosolic Sulfotransferase 1C2/SULT1C2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Cytosolic Sulfotransferase 1C2/SULT1C2
OTI4G1
Flow Cytometry
Affinity Purified, Conjugated
SULT1C2
Full length recombinant protein of human Cytosolic Sulfotransferase 1C2/SULT1C2 (NP_001047) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
cytosolic, 1C, member 1, EC 2.8.2, EC 2.8.2.-, EC 2.8.2.1, EC 2.8.2.2, humSULTC2, ST1C2, Sulfotransferase 1C1, sulfotransferase family, cytosolic, 1C, member 2, SULT1CNo.1
Mouse
IgG1
0.1 mL
Primary
6819.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit