Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Cytosolic Sulfotransferase 1E1/SULT1E1 Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
$1091.00
Specifications
Assay Range | 31.25 pg/mL |
---|---|
Assay Sensitivity | 31.25-2000 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB009667
|
Novus Biologicals™
NBP27931415PLATES |
15 Plates |
Each of 1 for $1,091.00
|
|
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Cytosolic Sulfotransferase 1E1/SULT1E1.Specifications
31.25 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
EC 2.8.2, EST-1, ESTMGC34459, estrone sulfotransferase, ST1E1, STEestrogen sulfotransferase, Sulfotransferase 1E1, sulfotransferase family 1E, estrogen-preferring, member 1, Sulfotransferase, estrogen-preferringEC 2.8.2.4 | |
SULT1E1 | |
Breast Cancer, Stem Cell Markers | |
Cytosolic Sulfotransferase 1E1/SULT1E1 |
31.25-2000 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Cytosolic Sulfotransferase 1E1/SULT1E1. | |
Sandwich ELISA | |
6783 | |
Rabbit | |
RUO | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 0.5 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 2 ng/mL is recommended. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title