Learn More
Cytosolic Sulfotransferase 2A1/SULT2A1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP299792AF647
Catalog No. NB062932
Add to Cart
Description
Description
Cytosolic Sulfotransferase 2A1/SULT2A1 Polyclonal specifically detects Cytosolic Sulfotransferase 2A1/SULT2A1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
Cytosolic Sulfotransferase 2A1/SULT2A1
Polyclonal
Western Blot, ELISA
alcohol/hydroxysteroid sulfotransferase, Dehydroepiandrosterone sulfotransferase, DHEAS, DHEA-STbile salt sulfotransferase, EC 2.8.2.14, HST, hSTa, Hydroxysteroid Sulfotransferase, ST2, ST2A1, ST2A3, STDbile-salt sulfotranasferase 2A1, Sulfotransferase 2A1, sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone(DHEA)-preferring, member 1
Produced in rabbits immunized with purified, recombinant Cytosolic Sulfotransferase 2A1/SULT2A1 (NP_003158.2; Ser 2-Glu 285).
0.1 mL
Primary
Human
Purified
Western Blot, ELISA
Alexa Fluor 647
50mM Sodium Borate
Rabbit
Affinity purified
RUO
6822
Store at 4°C in the dark.
IgG
Product Suggestions
Product Suggestions