Learn More
Cytosolic Sulfotransferase 2B1/SULT2B1 Rabbit anti-Human, DyLight 650, Clone: 7, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP289895C
Catalog No. NB109974
Add to cart
Description
Description
Cytosolic Sulfotransferase 2B1/SULT2B1 Monoclonal specifically detects Cytosolic Sulfotransferase 2B1/SULT2B1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
Cytosolic Sulfotransferase 2B1/SULT2B1
7
50 mM Sodium Borate
Alcohol sulfotransferase, EC 2.8.2, EC 2.8.2.2, HSST2sulfotransferase family cytosolic 2B member 1, Hydroxysteroid sulfotransferase 2, ST2B1, Sulfotransferase 2B1, sulfotransferase family, cytosolic, 2B, member 1
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cytosolic Sulfotransferase 2B1/SULT2B1 (NP_004596.2; Asp 2-Ser 365) .
Protein A purified
RUO
Primary
6820
Western Blot, ELISA
DyLight 650
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human