Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytosolic Sulfotransferase 2B1/SULT2B1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP154828
Description
Cytosolic Sulfotransferase 2B1/SULT2B1 Polyclonal specifically detects Cytosolic Sulfotransferase 2B1/SULT2B1 in Human samples. It is validated for Western Blot.Specifications
Cytosolic Sulfotransferase 2B1/SULT2B1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
O00204 | |
SULT2B1 | |
Synthetic peptides corresponding to SULT2B1(sulfotransferase family, cytosolic, 2B, member 1) The peptide sequence was selected from the C terminal of SULT2B1. Peptide sequence NTMSNYTLLPPSLLDHRRGAFLRKGVCGDWKNHFTVAQSEAFDRAYRKQM. | |
Affinity Purified | |
RUO | |
Primary | |
Rat: 86%; Porcine: 83%; . | |
Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
Alcohol sulfotransferase, EC 2.8.2, EC 2.8.2.2, HSST2sulfotransferase family cytosolic 2B member 1, Hydroxysteroid sulfotransferase 2, ST2B1, Sulfotransferase 2B1, sulfotransferase family, cytosolic, 2B, member 1 | |
Rabbit | |
41 kDa | |
100 μL | |
metabolism | |
6820 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title