Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Tocris Bioscience™ Cytosporone B Available on GSA/VA Contract for Federal Government customers only.

High affinity Nur77 agonist

$177.00 - $748.00

Specifications

Chemical Name or Material Ethyl 3,5-dihydroxy-2-(1-oxooctyl)-benzeneacetate
Molecular Formula C18H26O5
CAS 321661-62-5
Product Type Target: Nuclear Receptor Agonists
Formula Weight 322.4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
54-591-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5459/10
10mg Each for $177.00
N/A
54-595-0 View Documents Tocris Bioscience™
5459/50
50mg Each for $748.00
N/A
Description

Description

Naturally occurring Nur77 (NR4A1) agonist (Kd = 8.52 x 10-7 M). Induces apoptosis in gastric cancer cells in vitro. Increases blood glucose levels in fasting C57 mice and suppresses xenograft tumor growth. Also exhibits a potent antifibrotic effect in bleomycin-induced skin fibrosis in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Ethyl 3,5-dihydroxy-2-(1-oxooctyl)-benzeneacetate
321661-62-5
322.4
C18H26O5
Target: Nuclear Receptor Agonists
Safety and Handling

Safety and Handling

Recommended Storage : Store at −20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit