Learn More
BioVision IncSupplier Diversity Partner Bioluminescence Cytotoxicity Assay Kit: Bioluminescence Cytotoxicity Assay Kit: Highly Sensitive, High-Throughput Adaptable Bioluminescent Assay to Quantitate Cytotxicity. 500 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K312500
Cell death or cytotoxicity is classically evaluated by the quantification of plasma membrane damage. Adenylate kinase (AK) is a ubiquitous protein present in all eukaryotic and prokaryotic cells and rapidly release into culture medium upon damage of the plasma membrane. The Bioluminescence Cytotoxicity Assay Kit is based on the measurement of AK in a simple one-step procedure involving two chemical reactions. The first reaction is the conversion of ADP to ATP by adenylate kinase that was released from the damaged cells. The second reaction utilizes luciferase to catalyze the formation of light from ATP and luciferin, and the light can be measured using a luminometer or beta counter. The assay is highly sensitive and can be fully automatic for high throughput. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9686