CZC 24832, Tocris™

Selective inhibitor of PI 3-kinase γ

Manufacturer: R&D Systems 4674/50

Quantity 50mg

 View more versions of this product

Catalog No. 46-745-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C15H17FN6O2S
    Formula Weight 364.4
    Synonym 5-(2-Amino-8-fluoro[1,2,4]triazolo[1,5-a]pyridin-6-yl)-N-(1,1-dimethylethyl)-3-pyridinesulfonamide
    Chemical Name or Material CZC 24832

    Specifications

    Formula Weight 364.4
    Quantity 50mg
    Synonym 5-(2-Amino-8-fluoro[1,2,4]triazolo[1,5-a]pyridin-6-yl)-N-(1,1-dimethylethyl)-3-pyridinesulfonamide
    Molecular Formula C15H17FN6O2S
    CAS 1159824-67-5
    Recommended Storage Store at +4°C
    Assay Percent Range >98%
    Chemical Name or Material CZC 24832

    Store at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit