Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-10-Camphorsulfonyl chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
21286-54-4
C10H15ClO3S
250.74
MFCD00064156
BGABKEVTHIJBIW-GMSGAONNSA-N
d +-10-camphorsulfonyl chloride,1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonyl chloride,1s-+-camphor-10-sulfonyl chloride,d-2-oxobornane-10-sulphonyl chloride,1s-+-10-camphorsulfonyl chloride,d-+-10-camphorsulfonyl chloride,camphor-10-sulfonyl chloride,+-camphor-10-sulfonyl chloride,+-camphor-10-sulfonic acid chloride
88856
[(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonyl chloride
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)Cl)C
21286-54-4
C10H15ClO3S
250.74
MFCD00064156
BGABKEVTHIJBIW-GMSGAONNSA-N
d +-10-camphorsulfonyl chloride,1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonyl chloride,1s-+-camphor-10-sulfonyl chloride,d-2-oxobornane-10-sulphonyl chloride,1s-+-10-camphorsulfonyl chloride,d-+-10-camphorsulfonyl chloride,camphor-10-sulfonyl chloride,+-camphor-10-sulfonyl chloride,+-camphor-10-sulfonic acid chloride
88856
[(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonyl chloride
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)Cl)C
Infrared Spectrum Authentic
Quantity 25g
Color White to Beige
Specific Rotation + 33.00
Packaging Glass bottle
Melting Point 63.0°C to 68.0°C
Assay Percent Range 96% min. (ex Cl) (Argentometry)
Beilstein 11, 316
Fieser 01,109; 15,68
Formula Weight 250.74
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material D(+)-10-Camphorsulfonyl chloride, 97%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 244-314-8