Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-2-Allylglycine Hydrochloride 97.0+%, TCI America™

Click to view available options
Quantity:
200mg
1g
108412-04-0
C5H10ClNO2
151.59
MFCD06797041
DIDZZOWASZMNQW-PGMHMLKASA-N
(R)-2-Amino-4-pentenoic Acid Hydrochloride
44630066
(2R)-2-aminopent-4-enoic acid;hydrochloride
C=CCC(C(=O)O)N.Cl
108412-04-0
C5H10ClNO2
151.59
MFCD06797041
DIDZZOWASZMNQW-PGMHMLKASA-N
(R)-2-Amino-4-pentenoic Acid Hydrochloride
44630066
(2R)-2-aminopent-4-enoic acid;hydrochloride
C=CCC(C(=O)O)N.Cl
Color Undesignated
Quantity 1g
Melting Point 241°C
Formula Weight 151.59
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (T)
Chemical Name or Material D-2-Allylglycine Hydrochloride