Alfa Aesar™ D-2-Aminoadipic acid, 98%

Manufacturer: Alfa Aesar™ H2734503

Quantity 1g

 View more versions of this product

Catalog No. AAH2734503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7620-28-2
    Molecular Formula C6H11NO4
    Formula Weight 161.16
    MDL Number MFCD00063118
    Beilstein 1724347
    Synonym H-D-2-Aad-OH; (R)-2-Aminohexanedioic acid
    Chemical Name or Material D-2-Aminoadipic acid

    Specifications

    Formula Weight 161.16
    Molecular Formula C6H11NO4
    Melting Point 205°C
    Beilstein 1724347
    Quantity 1g
    MDL Number MFCD00063118
    CAS 7620-28-2
    Synonym H-D-2-Aad-OH; (R)-2-Aminohexanedioic acid
    Chemical Name or Material D-2-Aminoadipic acid
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit