Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-2-Aminoadipic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
7620-28-2
C6H11NO4
161.16
OYIFNHCXNCRBQI-SCSAIBSYSA-N
d-2-aminoadipic acid,r-2-aminohexanedioic acid,2r-2-aminohexanedioic acid,d-a-aminoadipic acid,d-,a-aminoadipic acid,r-2-aminoadipic acid,r-2-amino-hexanedioic acid,hexanedioic acid, 2-amino-,--and +,hexanedioic acid, 2-amino-, 2r,d-homoglutamic acid
165627
CHEBI:37025
(2R)-2-aminohexanedioic acid
C(CC(C(=O)O)N)CC(=O)O
7620-28-2
C6H11NO4
161.16
OYIFNHCXNCRBQI-SCSAIBSYSA-N
d-2-aminoadipic acid,r-2-aminohexanedioic acid,2r-2-aminohexanedioic acid,d-a-aminoadipic acid,d-,a-aminoadipic acid,r-2-aminoadipic acid,r-2-amino-hexanedioic acid,hexanedioic acid, 2-amino-,--and +,hexanedioic acid, 2-amino-, 2r,d-homoglutamic acid
165627
CHEBI:37025
(2R)-2-aminohexanedioic acid
C(CC(C(=O)O)N)CC(=O)O
Specific Rotation -20.5
Packaging Glass Bottle
Melting Point 186°C to 188°C
Color White
Elemental Analysis 43.60 to 45.84% C, 8.47 to 8.91% N
Quantity 1g
Infrared Spectrum Authentic
Assay Percent Range 98%
Formula Weight 161.16
Physical Form Powder or Crystalline Powder
Percent Purity 98%
Chemical Name or Material D-2-Aminoadipic acid
Show More Show Less