Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-(-)-2-Cyclohexylglycine, 95%

Click to view available options
Quantity:
1g
5g
25g
14328-52-0
C8H15NO2
157.213
MFCD01311678
WAMWSIDTKSNDCU-SSDOTTSWSA-N
d-cyclohexylglycine,d-alpha-cyclohexylglycine,r-2-amino-2-cyclohexylacetic acid,h-d-chg-oh,2-cyclohexyl-d-glycine,2r-amino cyclohexyl acetic acid,2r-2-amino-2-cyclohexylacetic acid,d-2-cyclohexylglycine,d-cyclohexyl glycine,h-cyclohexyl-d-gly-oh
736849
(2R)-2-amino-2-cyclohexylacetic acid
C1CCC(CC1)C(C(=O)O)N
14328-52-0
C8H15NO2
157.213
MFCD01311678
WAMWSIDTKSNDCU-SSDOTTSWSA-N
d-cyclohexylglycine,d-alpha-cyclohexylglycine,r-2-amino-2-cyclohexylacetic acid,h-d-chg-oh,2-cyclohexyl-d-glycine,2r-amino cyclohexyl acetic acid,2r-2-amino-2-cyclohexylacetic acid,d-2-cyclohexylglycine,d-cyclohexyl glycine,h-cyclohexyl-d-gly-oh
736849
(2R)-2-amino-2-cyclohexylacetic acid
C1CCC(CC1)C(C(=O)O)N
Beilstein 3196806
Formula Weight 157.21
Physical Form Solid
Percent Purity 95%
Quantity 1g
Chemical Name or Material D-(-)-2-Cyclohexylglycine

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures