Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D-2-Phosphoglyceric acid lithium salt, ≥65.0% (enzymatic); ≥95% (HPCE), MilliporeSigma™ Supelco™
$258.00 - $429.00
Specifications
CAS | 3443-57-0 |
---|---|
Type | Analytical Standard |
Linear Formula | C3H7O7 P · xLi+ |
Synonym | (2 R)-3-Hydroxy-2-(phosphonooxy)propanoic acid lithium salt; Lithium D-2-phosphoglycerate |
Formula Weight | 186.06 (free acid basis) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-101-3981
|
MilliporeSigma Supelco
7388510MG |
10 mg |
Each for $258.00
|
|
|||||
11-101-3982
|
MilliporeSigma Supelco
7388550MG |
50 mg |
Each for $429.00
|
|
|||||
Specifications
3443-57-0 | |
C3H7O7 P · xLi+ | |
186.06 (free acid basis) | |
Analytical Standard |
Analytical Standard | |
(2 R)-3-Hydroxy-2-(phosphonooxy)propanoic acid lithium salt; Lithium D-2-phosphoglycerate | |
≥65.0% (enzymatic); ≥95% (HPCE) |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title