Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips) Available on GSA/VA Contract for Federal Government customers only.
Achieve ultimate performance in automated pipetting with Thermo Scientific™ D.A.R.T.'s™.
Manufacturer: Thermo Scientific™ 5418
 View more versions of this product
Catalog No. 14-387-697
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    For Use With (Equipment) Thermo Scientific Matrix Hydra DT
    Filtered Yes
    Volume (Metric) 30μL,30µL
    Product Line D.A.R.T.'S
    Sterility Nonsterile
    Packaging 20 magazines of 384 tips/case,20 magazines of 384 tips/case
    Sterile No
    Features Extended length, filtered
    Achieve ultimate performance in automated pipetting with Thermo Scientific™ D.A.R.T.'s™. Unlike conventional automation tips, D.A.R.T.'s are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision. Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x3, PlateMate™ Plus Systems and Versette.
    Features:
    • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access.
    • Highly polished molds and cores ensure low liquid retention.
    • Small internal tip orifice provides better small volume droplet formation
    • Sterile, sterile filtered, wide-bore, and extended-length tip options are available
    • Tips are thoroughly inspected for proper formation and straightness
    • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing.
    • Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination.
    Includes: 20 magazines per case
    Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x3 and PlateMate™ Plus Systems.
  • Description & Specifications
Related Products
Product Content Feedback