Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Alanine, 99%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
Packaging:
Amber Glass Bottle
338-69-2
338-69-2
CAS Max % 1
Packaging Amber Glass Bottle
Percent Purity 0.99
Grade Reagent
Quantity 25 g