Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Alanine Methyl Ester Hydrochloride 97.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
14316-06-4
C4H10ClNO2
139.579
MFCD00066141
IYUKFAFDFHZKPI-AENDTGMFSA-N
d-alanine methyl ester hydrochloride,methyl d-alaninate hydrochloride,h-d-ala-ome.hcl,methyl d-alaninate hcl,methyl 2r-2-aminopropanoate hydrochloride,h-d-ala-ome hcl,r-methyl 2-aminopropanoate hydrochloride,d-alanine methylester hydrochloride,d-alanine, methyl ester, hydrochloride,pubchem10885
11182647
methyl (2R)-2-aminopropanoate;hydrochloride
CC(C(=O)OC)N.Cl
14316-06-4
C4H10ClNO2
139.579
MFCD00066141
IYUKFAFDFHZKPI-AENDTGMFSA-N
d-alanine methyl ester hydrochloride,methyl d-alaninate hydrochloride,h-d-ala-ome.hcl,methyl d-alaninate hcl,methyl 2r-2-aminopropanoate hydrochloride,h-d-ala-ome hcl,r-methyl 2-aminopropanoate hydrochloride,d-alanine methylester hydrochloride,d-alanine, methyl ester, hydrochloride,pubchem10885
11182647
methyl (2R)-2-aminopropanoate;hydrochloride
CC(C(=O)OC)N.Cl
Color White
Quantity 25g
Melting Point 108°C
Formula Weight 139.58
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (N,T)
Chemical Name or Material D-Alanine Methyl Ester Hydrochloride