Alfa Aesar™ D-allo-Isoleucine, 97%

Manufacturer: Alfa Aesar™ H2718903

Quantity 1g

 View more versions of this product

Catalog No. AAH2718903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1509-35-9
    Molecular Formula C6H13NO2
    Formula Weight 131.18
    MDL Number MFCD00066445
    Beilstein 1721794
    Synonym (2R,3S)-2-Amino-3-methylpentanoic acid; H-D-allo-Ile-OH
    Chemical Name or Material D-allo-Isoleucine

    Specifications

    Formula Weight 131.18
    Molecular Formula C6H13NO2
    MDL Number MFCD00066445
    CAS 1509-35-9
    Beilstein 1721794
    Melting Point ≈285°C
    Quantity 1g
    Optical Rotation −37° (c=1 in 5N HCl)
    RTECS Number BA2930000
    Synonym (2R,3S)-2-Amino-3-methylpentanoic acid; H-D-allo-Ile-OH
    Chemical Name or Material D-allo-Isoleucine
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit