Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-allo-Threonine, 99%, Thermo Scientific Chemicals

Catalog Number p-7029573
Click to view available options
Quantity:
250 mg
1 g
24830-94-2
C4H9NO3
119.12
MFCD00004526
AYFVYJQAPQTCCC-STHAYSLISA-N
d-allo-threonine, d-allothreonine, h-d-allo-thr-oh, d--allo-threonine, r-allothreonine, 2r,3r-2-amino-3-hydroxybutanoic acid, 2r,3r-allothreonine, r-allo-threonine, dl-allo-threonine, allothreonine, d
90624
CHEBI:32826
(2R,3R)-2-amino-3-hydroxybutanoic acid
C[C@H](O)[C@@H](N)C(O)=O
24830-94-2
C4H9NO3
119.12
MFCD00004526
AYFVYJQAPQTCCC-STHAYSLISA-N
d-allo-threonine, d-allothreonine, h-d-allo-thr-oh, d--allo-threonine, r-allothreonine, 2r,3r-2-amino-3-hydroxybutanoic acid, 2r,3r-allothreonine, r-allo-threonine, dl-allo-threonine, allothreonine, d
90624
CHEBI:32826
(2R,3R)-2-amino-3-hydroxybutanoic acid
C[C@H](O)[C@@H](N)C(O)=O

They are found as constituents of an increasing group of biologically active peptides. Stereocontrolled syntheses of chiral and racemic key intermediates to thienamycin from d-allo-threonine and trans-crotonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
They are found as constituents of an increasing group of biologically active peptides. Stereocontrolled syntheses of chiral and racemic key intermediates to thienamycin from d-allo-threonine and trans-crotonic acid.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 268°C (decomposition)
Beilstein 1721644
Solubility Information Soluble in water.
Formula Weight 119.12
Percent Purity 99%
Quantity 250 mg
Optical Rotation −8.7° (c=2 in Water)
Chemical Name or Material D-allo-Threonine
EINECSNumber 246-488-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit