Learn More
[D-Arg1, D-Phe5, D-Trp7,9, Leu11] Substance P, Alfa Aesar™
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar J66988MCR
 View more versions of this product
Catalog No. AAJ66988MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1516.85
1mg
D-Arg-Pro-Lys-Pro-D-Phe-Gln-D-Trp-Phe-D-Trp-Leu-Leu-NH2
-30° to -10°C
[D-Arg1, D-Phe5, D-Trp7,9, Leu11] Substance P
HPLC
Solid
RUO
Antagonist D