Learn More
[D-Arg0, Hyp3, Thi5,8, D-Phe7]-Adamantaneacetyl Bradykinin, Alfa Aesar™
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar J66709LB0
 View more versions of this product
Catalog No. AAJ66709LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1455.72
5mg
N-Adamantanecarbonyl-D-Arg-Arg-Pro-Hyp-Gly-Thi-Ser-D-Phe-Thi-Arg
-30° to -10°C
[D-Arg0, Hyp3, Thi5,8, D-Phe7]-Admantanecarbonyl Bradykinin
HPLC
Solid
RUO
[N-Admantanecarbonyl-D-Arg0-Hyp3, Thi, 5, 8, D-Phe7] Bradykinin