Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific™

Catalog Number AAH63795MD
Click to view available options
Quantity:
250 mg
1 g
5 g
64960-75-4
C8H15NO4
189.21
MFCD00077101
MXWMFBYWXMXRPD-UHFFFAOYNA-N
h-d-asp otbu-oh, d-aspartic acid 4-tert-butyl ester, r-2-amino-4-tert-butoxy-4-oxobutanoic acid, 2r-2-amino-4-tert-butoxy-4-oxobutanoic acid, d-aspartic acid beta-tert-butyl ester, d-aspartic acid 4-1,1-dimethylethyl ester, 2r-2-amino-4-tert-butoxy-4-oxo-butanoic acid, 2r-2-amino-4-2-methylpropan-2-yl oxy-4-oxobutanoic acid, 2r-2-azanyl-4-2-methylpropan-2-yl oxy-4-oxidanylidene-butanoic acid
7018828
CC(C)(C)OC(=O)CC(N)C(O)=O
64960-75-4
C8H15NO4
189.21
MFCD00077101
MXWMFBYWXMXRPD-UHFFFAOYNA-N
h-d-asp otbu-oh, d-aspartic acid 4-tert-butyl ester, r-2-amino-4-tert-butoxy-4-oxobutanoic acid, 2r-2-amino-4-tert-butoxy-4-oxobutanoic acid, d-aspartic acid beta-tert-butyl ester, d-aspartic acid 4-1,1-dimethylethyl ester, 2r-2-amino-4-tert-butoxy-4-oxo-butanoic acid, 2r-2-amino-4-2-methylpropan-2-yl oxy-4-oxobutanoic acid, 2r-2-azanyl-4-2-methylpropan-2-yl oxy-4-oxidanylidene-butanoic acid
7018828
CC(C)(C)OC(=O)CC(N)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
IUPAC Name 2-amino-4-(tert-butoxy)-4-oxobutanoic acid
Formula Weight 189.21
Percent Purity 98%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material D-Aspartic acid 4-tert-butyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit