Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ D-Aspartic acid dibenzyl ester p-toluenesulfonate, 95%

Click to view available options
Quantity:
25g
100g
4079-64-5
C25H27NO7S
485.551
MFCD00035102
HLMUYZYLPUHSNV-PKLMIRHRSA-N
h-d-asp obzl-obzl.tosoh,d-aspartic acid dibenzyl ester-p-toluenesulfonate,r-dibenzyl 2-aminosuccinate 4-methylbenzenesulfonate,d-aspartic acid dibenzyl ester tosylate,d-aspartic acid dibenzyl ester p-toluenesulfonate,d-aspartic acid dibenzyl ester 4-toluenesulfonate salt,d-aspdb tsoh,1,4-dibenzyl 2r-2-aminobutanedioate; para-toluene sulfonate,h-d-asp obzl-obzl tos,d,l-aspartic acid dibenzyl ester-p-toluenesulfonate
25188997
dibenzyl (2R)-2-aminobutanedioate;4-methylbenzenesulfonic acid
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CC(C(=O)OCC2=CC=CC=C2)N
4079-64-5
C25H27NO7S
485.551
MFCD00035102
HLMUYZYLPUHSNV-PKLMIRHRSA-N
h-d-asp obzl-obzl.tosoh,d-aspartic acid dibenzyl ester-p-toluenesulfonate,r-dibenzyl 2-aminosuccinate 4-methylbenzenesulfonate,d-aspartic acid dibenzyl ester tosylate,d-aspartic acid dibenzyl ester p-toluenesulfonate,d-aspartic acid dibenzyl ester 4-toluenesulfonate salt,d-aspdb tsoh,1,4-dibenzyl 2r-2-aminobutanedioate; para-toluene sulfonate,h-d-asp obzl-obzl tos,d,l-aspartic acid dibenzyl ester-p-toluenesulfonate
25188997
dibenzyl (2R)-2-aminobutanedioate;4-methylbenzenesulfonic acid
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CC(C(=O)OCC2=CC=CC=C2)N
Beilstein 6311519
Formula Weight 485.55
Physical Form Solid
Percent Purity 95%
Melting Point 155°C to 158°C
Quantity 100g
Chemical Name or Material D-Aspartic acid dibenzyl ester p-toluenesulfonate

storageNote1 : 2° to 8°C