Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Aspartic acid dibenzyl ester p-toluenesulfonate, 95%, Thermo Scientific™

Catalog Number AAH6547514
Click to view available options
Quantity:
25 g
100 g
4079-64-5
C25H27NO7S
485.551
MFCD00035102
HLMUYZYLPUHSNV-PKLMIRHRSA-N
h-d-asp obzl-obzl.tosoh, d-aspartic acid dibenzyl ester-p-toluenesulfonate, r-dibenzyl 2-aminosuccinate 4-methylbenzenesulfonate, d-aspartic acid dibenzyl ester tosylate, d-aspartic acid dibenzyl ester p-toluenesulfonate, d-aspartic acid dibenzyl ester 4-toluenesulfonate salt, d-aspdb tsoh, 1,4-dibenzyl 2r-2-aminobutanedioate; para-toluene sulfonate, h-d-asp obzl-obzl tos, d,l-aspartic acid dibenzyl ester-p-toluenesulfonate
25188997
dibenzyl (2R)-2-aminobutanedioate;4-methylbenzenesulfonic acid
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CC(C(=O)OCC2=CC=CC=C2)N
4079-64-5
C25H27NO7S
485.551
MFCD00035102
HLMUYZYLPUHSNV-PKLMIRHRSA-N
h-d-asp obzl-obzl.tosoh, d-aspartic acid dibenzyl ester-p-toluenesulfonate, r-dibenzyl 2-aminosuccinate 4-methylbenzenesulfonate, d-aspartic acid dibenzyl ester tosylate, d-aspartic acid dibenzyl ester p-toluenesulfonate, d-aspartic acid dibenzyl ester 4-toluenesulfonate salt, d-aspdb tsoh, 1,4-dibenzyl 2r-2-aminobutanedioate; para-toluene sulfonate, h-d-asp obzl-obzl tos, d,l-aspartic acid dibenzyl ester-p-toluenesulfonate
25188997
dibenzyl (2R)-2-aminobutanedioate;4-methylbenzenesulfonic acid
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CC(C(=O)OCC2=CC=CC=C2)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 155°C to 158°C
Beilstein 6311519
Formula Weight 485.55
Percent Purity 95%
Quantity 25 g
Physical Form Solid
Chemical Name or Material D-Aspartic acid dibenzyl ester p-toluenesulfonate
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit