Alfa Aesar™ D-beta-Homoproline hydrochloride, 95%

Manufacturer: Alfa Aesar™ H5208503

Quantity 1g

 View more versions of this product

Catalog No. AAH5208503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 439918-59-9
    Molecular Formula C6H11NO2·HCl
    Formula Weight 165.62
    MDL Number MFCD06202401
    Synonym (R)-2-(2-Pyrrolidinyl)acetic acid hydrochloride
    Chemical Name or Material D-beta-Homoproline hydrochloride

    Specifications

    Formula Weight 165.62
    Molecular Formula C6H11NO2·HCl
    MDL Number MFCD06202401
    CAS 439918-59-9
    Synonym (R)-2-(2-Pyrrolidinyl)acetic acid hydrochloride
    Melting Point 195° to 197°C
    Quantity 1g
    Optical Rotation −36° (c=1 in Water)
    Chemical Name or Material D-beta-Homoproline hydrochloride
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit