Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D and C Green No. 6, 96%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
128-80-3
C28H22N2O2
418.50
TVRGPOFMYCMNRB-UHFFFAOYSA-N
1,4-bis[(4-methylphenyl)amino]-9,10-dihydroanthracene-9,10-dione
CC1=CC=C(NC2=CC=C(NC3=CC=C(C)C=C3)C3=C2C(=O)C2=CC=CC=C2C3=O)C=C1
128-80-3
C28H22N2O2
418.50
TVRGPOFMYCMNRB-UHFFFAOYSA-N
1,4-bis[(4-methylphenyl)amino]-9,10-dihydroanthracene-9,10-dione
CC1=CC=C(NC2=CC=C(NC3=CC=C(C)C=C3)C3=C2C(=O)C2=CC=CC=C2C3=O)C=C1

D1126, 128-80-3, C28H22N2O2

Spectrum™ D and C Green No. 6
CAS Max % 100%
Arsenic (As) 3 ppm
Percent Purity 96%
Lead (Pb) 20 ppm
Mercury (Hg) 1 ppm
Packaging Amber Glass Bottle
Quantity 500 g