Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D and C Red No. 17, 90%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
85-86-9
C22H16N4O
352.40
HTPQPMPFXUWUOT-SRURNFRUSA-N
(1Z)-1-{2-[4-(2-phenyldiazen-1-yl)phenyl]hydrazin-1-ylidene}-1,2-dihydronaphthalen-2-one
O=C1C=CC2=CC=CC=C2\C1=N\NC1=CC=C(C=C1)N=NC1=CC=CC=C1
85-86-9
C22H16N4O
352.40
HTPQPMPFXUWUOT-SRURNFRUSA-N
(1Z)-1-{2-[4-(2-phenyldiazen-1-yl)phenyl]hydrazin-1-ylidene}-1,2-dihydronaphthalen-2-one
O=C1C=CC2=CC=CC=C2\C1=N\NC1=CC=C(C=C1)N=NC1=CC=CC=C1

D1132, 85-86-9, C22H16N4O

Spectrum™ D and C Red No. 17 - is a certified oil soluble cosmetic color used in drug and cosmetic products such as creams, lotions, gels, hair products, bath products and soaps to produce a yellowish red. Small quantities are required since it is very concentrated.
CAS Max % 100%
Arsenic (As) 3 ppm
Percent Purity 90%
Lead (Pb) 20 ppm
Mercury (Hg) 1 ppm
Packaging Amber Glass Bottle
Quantity 500 g