Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D and C Red No. 33, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
3567-66-6
3567-66-6

DC145, 3567-66-6, C16H11N3O7S2Na2

Spectrum™ D and C Red No. 33
CAS Max % 100%
Arsenic (As) 3 ppm
Lead (Pb) 20 ppm
Mercury (Hg) 1 ppm
Packaging Amber Glass Bottle
Quantity 100 g